missing translation for 'onlineSavingsMsg'
Learn More

GATSL3 Antibody [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30498642 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498642 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498642 Supplier Novus Biologicals Supplier No. NBP337902UV
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GATSL3 Polyclonal antibody specifically detects GATSL3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATSL3
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias GATS protein-like 3, GATS-like protein 3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-329 of human GATSL3 (NP_001032755.1).,, Sequence:, MELHILEHRVRVLSVARPGLWLYTHPLIKLLFLPRRSRCKFFSLTETPEDYTLMVDEEGFKELPPSEFLQVAEATWLVLNVSSHSGAAVQAAGVTKIARSVIAPLAEHHVSVLMLSTYQTDFILVREQDLSVVIHTLAQEFDIYREVGGEPVPVTRDDSSNGFPRTQHGPSPTVHPIQSPQNRFCVLTLDPETLPAIATTLIDVLFYSHSTPKEAASSSPEPSSITFFAFSLIEGYISIVMDAETQKKFPSDLLLTSSSGELWRMVRIGGQPLGFDECGIVAQIAGPLAAADISAYYISTFNFDHALVPEDGIGSVIEVLQRRQEGLAS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 652968
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.