missing translation for 'onlineSavingsMsg'
Learn More

MEIS1 Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30498516 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498516 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498516

Brand: Novus Biologicals NBP335814PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEIS1 Polyclonal antibody specifically detects MEIS1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIS1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-390 of human MEIS1 (NP_002389.1).,, Sequence:, PSEEQKKQLAQDTGLTILQVNNWFINARRRIVQPMIDQSNRAVSQGTPYNPDGQPMGGFVMDGQQHMGIRAPGPMSGMGMNMGMEGQWHYM
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4211
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.