missing translation for 'onlineSavingsMsg'
Learn More

Cullin 1 Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30498383 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498383 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498383

Brand: Novus Biologicals NBP337968PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cullin 1 Polyclonal antibody specifically detects Cullin 1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias CUL-1, cullin 1, cullin-1, MGC149834, MGC149835
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 435-593 of human Cullin 1 (NP_003583.2).,, Sequence:, KNPEEAELEDTLNQVMVVFKYIEDKDVFQKFYAKMLAKRLVHQNSASDDAEASMISKLKQACGFEYTSKLQRMFQDIGVSKDLNEQFKKHLTNSEPLDLDFSIQVLSSGSWPFQQSCTFALPSELERSYQRFTAFYASRHSGRKLTWLYQLSKGELVTN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8454
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.