Learn More
Sampling Systems™ 304 Stainless Steel Storage Containers without Toggle Clamps
Product Code. 15819681 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
20 L
25 L
30 L
50 L
75 L
100 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15819681 30 L Each
15899671 20 L Each
15809681 25 L Each
15829681 50 L Each
15839681 75 L Each
15889671 100 L Each
6 options
Product Code. 15819681
Brand: Sampling Systems™ A45430
These 304 Stainless Steel Storage Containers are an excellent option for multi-purpose storage in the laboratory.
  • Manufactured from durable 304 grade stainless steel
  • Crevice-free interior
  • Close fitting recessed lid
  • Each with two sturdy handles
  • Flat bottom with chime ring to give added protection
  • Light duty construction
  • Available in a wide range of capacities to fit your specific needs
Specifications
Closure Included Yes
Product Type Storage container
Closure Type Recessed Lid with Handle
Capacity (Metric) 30 L
For Use With (Application) Multi-purpose Storage
Material Stainless Steel
Height (Metric) 445 mm
Diameter (Metric) Outer 300 mm