Learn More
Sampling Systems™ 304 Stainless Steel Micro Spatula
Product Code. 12756267 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
175 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
12756267 175 mm Each
1 options
Product Code. 12756267
Brand: Sampling Systems™ A10020
Only null left
Add to basket
Add to basket
Sampling Systems™ 304 Stainless Steel Micro Spatula is ideal for sampling or dispensing small volumes of powder.
  • Stainless steel
  • For powder sampling
  • Crevice free
  • Bright polish finish
Specifications
Product Type Micro Spatula
Length (Metric) Blade 40 mm
Width (Metric) Blade 12 mm
Material Stainless Steel (304)
Length (Metric) 175 mm