Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Specifications
| Material | Nickel-plated Brass (Hub), Stainless Steel (304) |
| Needle Gauge | 19 G |
| Diameter (Metric) Inner | 0.69 mm |
| Diameter (Metric) Outer | 1.07 mm |
| Needle Point Style | 5 or AS |
| For Use With (Equipment) | TLL Syringes and LT or TLL Connectors |