Learn More
Nickel Electro™ Stainless Steel Curved Half Moon End Hayman Spatula
Shop All Nickel-Electro ProductsDescription
- Stainless steel
- Hayman Spatula
- Non-magnetic
- Bright finish
- This Spatula has a 3mm wide curved half moon end at the tip and approximately 13 mm long
- One end is sharpened at the tip for cutting, separating or handling micro samples
- The other end has a flat blade for handling small samples
Specifications
Specifications
| Quantity | 1 Ea. |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Product Type | Spatula |
| Finish | Bright |
| For Use With (Application) | Sterile Applications |
| Temperature (Metric) Max | -196°C to +550°C |
| Handle Material | Stainless Steel |
| Material | Stainless Steel |
| Length (Metric) | 165 mm |