Learn More
TubeWriter 300 - UV Curable Ink
Product Code. 30067437
Change view
Click to view available options
Product Type:
Ink Roll Replacement
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30067437 Ink Roll Replacement Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30067437 Supplier TubeWriter Supplier No. TWUVB3001
  • ONLINE EXCLUSIVE PRICE
    16430.00 NOK
    14184.40 NOK / Pack of 1
    Save 2245.60 NOK
    13% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Replacement UV curable ink
  • Ink prints and labels directly on curved plastics and glass surfaces - any labware
  • UV curation ensures printing stands up to harsh laboratory conditions
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Resistant to chemical solvents
  • Disposable cartridges with low replacement costs
  • No time-consuming sticky labels or handwriting required
Specifications
Product Type Ink Roll Replacement