Learn More
TubeWriter 300 - Labeling System
Product Code. 30067427
Change view
Click to view available options
Includes:
2 Labeling Racks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
30067427 2 Labeling Racks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30067427 Supplier TubeWriter Supplier No. TW300A
  • ONLINE EXCLUSIVE PRICE
    563200.00 NOK
    486322.19 NOK / Pack of 1
    Save 76877.81 NOK
    13% Off
View all promotions
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Base Labeling System - Includes 2 Standard Racks
  • Full-featured instrument for printing labels directly on any labware or curved surface
  • Compact footprint for labs with tighter bench space - 20 in. x 18 in. x 18 in. (WxLxH)
  • Designed for the needs of scaling labs
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications
Product Type Labeling System
Includes 2 Labeling Racks