Learn More
Lenz Laborglasintrumente 3-Way Stopcock with Solid Glass Plug
Product Code. p-7222456
Change view
Click to view available options
Plug Bore:
1.5 mm
2.5 mm
6 mm
Unit Size:
Each
Product Code. Plug Bore unitSize
10120251 1.5 mm Each
10291801 2.5 mm Each
10251901 6 mm Each
3 options
Product Code. 10120251
Brand: Lenz Laborglasintrumente 02501401
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12554
  • T-bore, with solid glass plug and screwthread retaining device
  • Material: borosilicate glass 3.3
  • Leakage rate to DIN 12540-1
Specifications
Plug Bore 1.5 mm
Material Borosilicate Glass (3.3)