missing translation for 'onlineSavingsMsg'
Learn More

3-Vinylbenzoic acid, 96%

Product Code. 15405297
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15405297

Brand: Thermo Scientific Alfa Aesar H32556.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28447-20-3 | C9H8O2 | 148.161 g/mol

Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, ultraviolet radiations, free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28447-20-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00157038
InChI Key VWXZFDWVWMQRQR-UHFFFAOYSA-N
PubChem CID 4438231
IUPAC Name 3-ethenylbenzoic acid
SMILES C=CC1=CC(=CC=C1)C(=O)O

Specifications

Melting Point 95°C
Quantity 1 g
Synonym 3-vinylbenzoic acid, 3-vinyl benzoic acid, 3-ethenyl-benzoic acid, benzoic acid,3-ethenyl, benzoic acid, 3-ethenyl, benzoic acid, 3-ethenyl-9ci
Formula Weight 148.16
Percent Purity 96%
Chemical Name or Material 3-Vinylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.