missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilyl-2-oxazolidinone, Thermo Scientific™

Product Code. p-7051442
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11359577 1 g 1g
11379577 5 g 5g
11369577 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11359577

Brand: Thermo Scientific Alfa Aesar 041209.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43112-38-5
Molecular Formula C6H13NO2Si
Molecular Weight (g/mol) 159.26
MDL Number MFCD00010197
InChI Key AUKCYOUETBBMFV-UHFFFAOYSA-N
Synonym 3-trimethylsilyl-2-oxazolidinone, 3-trimethylsilyl oxazolidin-2-one, n-trimethylsilyl-2-oxazolidinone, 3-trimethylsilyl-2-oxazolidone, acmc-20amyd, pubchem24175, 2-oxazolidinone,3-trimethylsilyl, trimethyl 2-oxo-3-oxazolidinyl silane, 2-oxo-3-trimethylsilyl-1,3-oxazolidine
PubChem CID 123514
IUPAC Name 3-trimethylsilyl-1,3-oxazolidin-2-one
SMILES C[Si](C)(C)N1CCOC1=O

Specifications

Density 1.013 g/mL
Boiling Point 99°C to 100°C (6 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.454
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 159.26
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.