Learn More
3-Trimethylsilyl-2-oxazolidinone, Thermo Scientific™
Product Code. 11359577
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11359577
Brand: Thermo Scientific Alfa Aesar 041209.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 43112-38-5
Molecular Formula C6H13NO2Si
Molecular Weight (g/mol) 159.26
MDL Number MFCD00010197
InChI Key AUKCYOUETBBMFV-UHFFFAOYSA-N
Synonym 3-trimethylsilyl-2-oxazolidinone, 3-trimethylsilyl oxazolidin-2-one, n-trimethylsilyl-2-oxazolidinone, 3-trimethylsilyl-2-oxazolidone, acmc-20amyd, pubchem24175, 2-oxazolidinone,3-trimethylsilyl, trimethyl 2-oxo-3-oxazolidinyl silane, 2-oxo-3-trimethylsilyl-1,3-oxazolidine
PubChem CID 123514
IUPAC Name 3-trimethylsilyl-1,3-oxazolidin-2-one
SMILES C[Si](C)(C)N1CCOC1=O
Specifications
Density 1.013 g/mL
Boiling Point 99°C to 100°C (6 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.454
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 159.26
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone