Learn More
Ismatec™ 3-Stop Tubing, Tygon E-LFL
Product Code. 15783329
Click to view available options
Tubing Size:
I.D. 0.27 mm
I.D. 0.38 mm
I.D. 0.76 mm
I.D. 1.14 mm
I.D. 1.25 mm
I.D. 1.37 mm
I.D. 1.60 mm
I.D. 2.29 mm
Unit Size:
Pack of 12
Product Code. 15783329
Brand: Ismatec™ SCE0405
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Durable, chemically resistant pump tubing available in a broad range of sizes.
  • DEHP free.
  • Longest service life of any clear Tygon tubing material.
  • Excellent choice where transparency and good chemical resistance is needed.
  • Broad chemical resistance.
  • Tasteless.
  • Extremely low particulate spallation.
  • Meets USP Class VI and FDA criteria.
  • Non-Aging.
Specifications
Certifications/Compliance FDA 21 CFR 175.300; US Pharmacopoea Class VI
Color Code Gray-Gray
Autoclavable Autoclavable
Diameter (Metric) Inner 1.25 mm
Material Tygon E-LFL
Overall Length (Metric) 400 mm
For Use With (Application) Pumps that incorporate cassettes into the pumphead design.
Min. Operating Temperature (Metric) −50°C
Packaging Quantity 12/Pk
Resistance Properties Acids: Fair, Alkaline solutions: Fair, Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent Sterile media: Limited
Tubing Size I.D. 1.25 mm
Sterilization Method Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky. Gas sterilization with ethylene oxide. Not recommended for sterilization with radiation.
Color Transparent
Max. Operating Temperature (Metric) 74°C