Learn More
3-Pyridinecarbaldehyde, Sigma-Aldrich™
Specifications
Specifications
| CAS | 500-22-1 |
| Melting Point | 7°C |
| Density | 1.14 g/cm3 at 20°C |
| Molecular Formula | 3-(CHO)C5H4N |
| MDL Number | MFCD00006382 |
| Quantity | 50 mL |
| Synonym | 3-Pyridinecarbaldehyde, Nicotinaldehyde, Pyridine-3-aldehyde, 3-Pyridinecarbaldehyde |
| Molecular Weight (g/mol) | 107.11 |
| Grade | Research |