Learn More
3-Propionylpyridine, 98+%
Product Code. 11372227
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372227 5 g 5g
11362227 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11372227 Supplier Thermo Scientific Alfa Aesar Supplier No. L09711.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 1570-48-5 | C8H9NO | 135.166 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1570-48-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00023589
InChI Key VDNKJMUNLKAGAM-UHFFFAOYSA-N
Synonym 3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
PubChem CID 74074
IUPAC Name 1-pyridin-3-ylpropan-1-one
SMILES CCC(=O)C1=CN=CC=C1
Specifications
Density 1.114
Boiling Point 109°C to 112°C (10 mmHg)
Refractive Index 1.524
Quantity 5 g
Beilstein 111365
Formula Weight 135.17
Percent Purity ≥98%
Chemical Name or Material 3-Propionylpyridine