Description
- Constructed from a combination of pleated clear, polyethylene apertured film and PTFE (polytetrafluoroethylene), with soft tie strings
- Manufactured in systems complying ISO 13485:2016 requirements
- Certified Sterile
Specifications
Specifications
| Product Type | Cleanroom Face Mask |
| Color | White |
| Sterility | Sterile |
| Style | Pleated with Ties |
| Certifications/Compliance | The masks are rigorously tested and manufactured in an ISO 13485 certified environment. The tests are validated by an independent control laboratory. This product is not a medical device in context of Regulation (EU) 2017/745/Directive 93/42/EEC nor personal protective equipment in context of Regulation (EU) 2016/425. Each batch is sterilized following ISO 11137 and substantiate a sterility assurance level SAL 10−6 according to ISO 11137-2:2013. |
| Disposable | Single-use |
| For Use With (Application) | Industrial Use Only |
| Length (Metric) | 23 cm |