missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)benzaldehyde, 97%, Thermo Scientific™

Catalog Number 15417487
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
401892-80-6
C7H5F5OS
232.168
MFCD03788517
APFMXGBPENUHDD-UHFFFAOYSA-N
3-pentafluorosulfanyl benzaldehyde, 3-pentafluorothio benzaldehyde, 3-pentafluoro-??-sulfanyl benzaldehyde, pubchem8517, 3-sulphurpentafluorobenzaldehyde, 3-pentafluorosulfur benzaldehyde, 3-formylphenyl pentafluorosulfur vi, 3-pentafluoro-$l^ 6-sulfanyl benzaldehyde, 3-pentakis fluoranyl-$l^ 6-sulfanyl benzaldehyde
2779205
3-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)C=O
This item is not returnable. View return policy
401892-80-6
C7H5F5OS
232.168
MFCD03788517
APFMXGBPENUHDD-UHFFFAOYSA-N
3-pentafluorosulfanyl benzaldehyde, 3-pentafluorothio benzaldehyde, 3-pentafluoro-??-sulfanyl benzaldehyde, pubchem8517, 3-sulphurpentafluorobenzaldehyde, 3-pentafluorosulfur benzaldehyde, 3-formylphenyl pentafluorosulfur vi, 3-pentafluoro-$l^ 6-sulfanyl benzaldehyde, 3-pentakis fluoranyl-$l^ 6-sulfanyl benzaldehyde
2779205
3-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 232.17
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'