Learn More
TERUMO 3-Part Syringes with Needle
Product Code. 17865522
Change view
Click to view available options
Needle Gauge:
20 G
21 G
22 G
23 G
25 G
27 G
Length (Metric):
13 mm
16 mm
25 mm
28 mm
32 mm
38 mm
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
Unit Size:
Pack of 100
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (Metric) Volume (Metric) Unit Size
17865522 27 G 13 mm 1 mL Pack of 100
17735039 21 G 16 mm 3 mL Pack of 100
17745039 21 G 25 mm 3 mL Pack of 100
17755039 21 G 38 mm 3 mL Pack of 100
17765039 22 G 28 mm 3 mL Pack of 100
17775039 23 G 25 mm 3 mL Pack of 100
17785039 23 G 32 mm 3 mL Pack of 100
17795039 25 G 16 mm 3 mL Pack of 100
17172714 21 G 38 mm 5 mL Pack of 100
17182714 22 G 38 mm 5 mL Pack of 100
17192714 23 G 32 mm 5 mL Pack of 100
17112724 20 G 38 mm 10 mL Pack of 100
17132724 22 G 38 mm 10 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 17865522 Supplier TERUMO Supplier No. 8MDSS01S2713E
  • ONLINE EXCLUSIVE PRICE
    183.75 NOK
    125.39 NOK / Pack of 100
    Save 58.36 NOK
    31% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Single use pre-connected with a sterile hypodermic needle
  • Every single detail is the result of Terumo long time experience in this field which is appreciated by users throughout the world
  • Sterile 3-part syringe for single use pre-connected with a sterile hypodermic needle (1 - 10 mL, 21 - 27 G)
  • Ultra-sharp bevel design for easy penetration
  • High quality silicone coating for smooth needle movement
  • Firm double end stop to prevent accidental removal of the piston and spilling of medication
  • Double contact gasket prevents leakage
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Polypropylene (Plunger)
Needle Gauge 27 G
Latex Free Yes
Length (Metric) 13 mm
Product Type Syringe With Needle
Disposable Single-use
Volume (Metric) 1 mL
Connection Type Luer Slip
Graduated Yes