Learn More
3-Nitrophthalic Anhydride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 641-70-3 |
| Melting Point | 160°C to 164°C |
| MDL Number | MFCD00005921 |
| Quantity | 25 g |
| Synonym | 3-Nitrophthalic anhydride, 3-Nitrophthalic anhydride |
| Molecular Weight (g/mol) | 193.11 |
| Grade | Research |