missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenol, Supelco™

Product Code. 15817530
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817530 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817530 Supplier Supelco Supplier No. 1067940025
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 95°C to 98°C
Boiling Point 194°C/93 hPa
CAS 554-84-7
MDL Number MFCD00007240
Density 1.49 g/cm3 at 20°C
Synonym 3-Nitrophenol, m-Nitrophenol, 3-Nitrophenol
Quantity 25 g
Molecular Weight (g/mol) 139.11
Grade Research
Chemical Name or Material 3-Nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.