Learn More
3-Nitrobenzoic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 121-92-6 |
| Melting Point | 139°C to 142°C |
| Density | 1.49 g/cm3 at 20°C |
| Molecular Formula | 3-(NO2)C6H4COOH |
| MDL Number | MFCD00007251 |
| Quantity | 250 g |
| Synonym | 3-Nitrobenzoic acid, 3-Nitrobenzoic acid |
| Molecular Weight (g/mol) | 167.12 |
| Grade | Research |