Learn More
Lenz Laborglasintrumente 3-Neck Adapters
Product Code. p-7222469
Change view
Click to view available options
Standard Taper Joint No.:
Cone 14/23 NS, Sockets 14/23 NS
Cone 24/29 NS, Sockets 19/26 NS
Cone 24/29 NS, Sockets 24/29 NS
Unit Size:
Each
Product Code. Standard Taper Joint No. unitSize
10376261 Cone 14/23 NS, Sockets 14/23 NS Each
10438982 Cone 24/29 NS, Sockets 19/26 NS Each
10448982 Cone 24/29 NS, Sockets 24/29 NS Each
3 options
Product Code. 10376261
Brand: Lenz Laborglasintrumente 05015011
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With 3 NS sockets and 1 NS cone
  • With two parallel necks and one angled neck (45°)
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Standard Taper Joint No. Cone 14/23 NS, Sockets 14/23 NS