Learn More
Lenz Laborglasintrumente 3-Neck Adapters
Product Code. 10448982
Change view
Click to view available options
Standard Taper Joint No.:
Cone 14/23 NS, Sockets 14/23 NS
Cone 24/29 NS, Sockets 19/26 NS
Cone 24/29 NS, Sockets 24/29 NS
Unit Size:
Each
Product Code. Standard Taper Joint No. unitSize
10448982 Cone 24/29 NS, Sockets 24/29 NS Each
10376261 Cone 14/23 NS, Sockets 14/23 NS Each
10438982 Cone 24/29 NS, Sockets 19/26 NS Each
3 options
Product Code. 10448982
Brand: Lenz Laborglasintrumente 05015033
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With 3 NS sockets and 1 NS cone
  • With two parallel necks and one angled neck (45°)
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Standard Taper Joint No. Cone 24/29 NS, Sockets 24/29 NS