Learn More
Lenz Laborglasintrumente 3-Neck Adapters
Description
- With 3 NS sockets and 1 NS cone
- With two parallel necks and one angled neck (45°)
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Standard Taper Joint No. | Cone 14/23 NS, Sockets 14/23 NS |