Learn More
3-N-Boc-5'-O-DMT-3'-O-Nosyl Thymidine, TRC
Product Code. 30474152
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474152 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30474152 Supplier Toronto Research Chemicals Supplier No. TRCN212160100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-N-Boc-5'-O-dimethoxytrityl-3'-O-nosylthymidine
CAS 444717-23-1
Molecular Formula C42 H43 N3 O13 S
Quantity 100 mg
Synonym 1(2H)-Pyrimidinecarboxylic acid, 3-[5-O-[bis(4-methoxyphenyl)phenylmethyl]-2-deoxy-3-O-[(4-nitrophenyl)sulfonyl]-β-D-threo-pentofuranosyl]-3,6-dihydro-5-methyl-2,6-dioxo-, 1,1-dimethylethyl ester (9CI, ACI), 1,1-Dimethylethyl 3-[5-O-[bis(4-methoxyphenyl)p
InChI Formula InChI=1S/C42H43N3O13S/c1-27-25-43(39(47)44(38(27)46)40(48)58-41(2,3)4)37-24-35(56-33-20-22-34(23-21-33)59(51,52)45(49)50)36(57-37)26-55-42(28-10-8-7-9-11-28,29-12-16-31(53-5)17-13-29)30-14-18-32(54-6)19-15-30/h7-23,25,35-37H,24,26H2,1-6H3/t35-,36+,37+/m0/
SMILES COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2Oc3ccc(cc3)S(=O)(=O)[N+](=O)[O-])N4C=C(C)C(=O)N(C(=O)OC(C)(C)C)C4=O)(c5ccccc5)c6ccc(OC)cc6
IUPAC Name tert-butyl 3-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-(4-nitrosulfonylphenoxy)oxolan-2-yl]-5-methyl-2,6-dioxopyrimidine-1-carboxylate
Molecular Weight (g/mol) 829.868
Formula Weight 829.252