missing translation for 'onlineSavingsMsg'
Learn More

3-Methyltoxoflavin, MedChemExpress

Product Code. p-200097705
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477127

Brand: Medchem Express HY1111175mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Methyltoxoflavin is a potent Protein disulfide isomerase (PDI) inhibitor, with an IC50 of 170 nM.

3-Methyltoxoflavin is a potent Protein disulfide isomerase (PDI) inhibitor, with an IC50 of 170 nM.

Specifications

Chemical Name or Material 3-Methyltoxoflavin
Physical Form Solid
CAS 32502-62-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H9N5O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 150 mg/mL (723.97 mM; Need ultrasonic)
SMILES CC1=NN(C)C(C2=N1)=NC(N(C)C2=O)=O
Molecular Weight (g/mol) 207.19
Formula Weight 207.19
Percent Purity 99.64%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.