Learn More
3-Methyltoxoflavin, MedChemExpress
Product Code. 16477127
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16477127
Brand: Medchem Express HY1111175mg
MedChemExpress 3-Methyltoxoflavin is a potent Protein disulfide isomerase (PDI) inhibitor, with an IC50 of 170 nM.
3-Methyltoxoflavin is a potent Protein disulfide isomerase (PDI) inhibitor, with an IC50 of 170 nM.
Specifications
Chemical Name or Material 3-Methyltoxoflavin
Physical Form Solid
CAS 32502-62-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H9N5O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 150 mg/mL (723.97 mM; Need ultrasonic)
SMILES CC1=NN(C)C(C2=N1)=NC(N(C)C2=O)=O
Molecular Weight (g/mol) 207.19
Formula Weight 207.19
Percent Purity 99.64%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.