Learn More
3'-Methylbiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™
Catalog Number p-4812963
Click to view available options
Quantity:
1 g
5 g
186551-47-3
C13H11ClO2S
266.75
OHVJVSGIYVAZFD-UHFFFAOYSA-N
3'-methyl-biphenyl-4-sulfonyl chloride, 3'-methylbiphenyl-4-sulfonyl chloride, 3'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methyl, 4-3-methylphenyl benzenesulfonyl chloride, 3/'-methylbiphenyl-4-sulfonyl chloride, 3'-methyl 1,1'-biphenyl-4-sulfonyl chloride
10754271
4-(3-methylphenyl)benzenesulfonyl chloride
CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
186551-47-3
C13H11ClO2S
266.75
OHVJVSGIYVAZFD-UHFFFAOYSA-N
3'-methyl-biphenyl-4-sulfonyl chloride, 3'-methylbiphenyl-4-sulfonyl chloride, 3'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methyl, 4-3-methylphenyl benzenesulfonyl chloride, 3/'-methylbiphenyl-4-sulfonyl chloride, 3'-methyl 1,1'-biphenyl-4-sulfonyl chloride
10754271
4-(3-methylphenyl)benzenesulfonyl chloride
CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 266.75
Percent Purity 97%
Chemical Name or Material 3'-Methylbiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 3'-Methylbiphenyl-4-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.