missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylbiphenyl-4-carboxaldehyde, 96%, Thermo Scientific™

Catalog Number 15487087
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
400744-83-4
C14H12O
196.249
MFCD03424618
LUJIYJCRJKYCRK-UHFFFAOYSA-N
2759813
4-(3-methylphenyl)benzaldehyde
CC1=CC=CC(=C1)C2=CC=C(C=C2)C=O
This item is not returnable. View return policy
400744-83-4
C14H12O
196.249
MFCD03424618
LUJIYJCRJKYCRK-UHFFFAOYSA-N
2759813
4-(3-methylphenyl)benzaldehyde
CC1=CC=CC(=C1)C2=CC=C(C=C2)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 45°C to 49°C
Flash Point >110°C (230°F)
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 4-3-methylphenyl benzaldehyde, 3'-methyl-1,1'-biphenyl-4-carbaldehyde, 3'-methylbiphenyl-4-carbaldehyde, 3'-methyl-biphenyl-4-carbaldehyde, 3'-methylbiphenyl-4-carboxaldehyde, 3'-methyl-biphenyl-4-carboxaldehyde, 1,1'-biphenyl-4-carboxaldehyde,3'-methyl, 1,1'-biphenyl-4-carboxaldehyde, 3'-methyl, pubchem10294, 4-m-tolyl benzaldehyde
Formula Weight 196.25
Percent Purity 96%
Chemical Name or Material 3'-Methylbiphenyl-4-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'