missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzoic Acid, Sigma-Aldrich™

Product Code. 15861561
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861561 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15861561 Supplier Sigma Aldrich Supplier No. 8219020250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-04-7
Grade Research
Molecular Formula 3-(CH3)C6H4COOH
Molecular Weight (g/mol) 136.15

Specifications

Melting Point 108°C to 111°C
Boiling Point 263°C/1013 hPa
MDL Number MFCD00002523
Quantity 250 g
Synonym 3-Methylbenzoic acid, m-Toluic acid, 3-Methylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.