missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-decanol, erythro + threo, 97%, Thermo Scientific™

Catalog Number 11490521
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
55816-17-6
C11H24O
172.312
MFCD00046702
SBMFCRFCWMMVQC-UHFFFAOYSA-N
3-methyl-4-decanol, 4-decanol, 3-methyl, 2-butyl n-hexyl carbinol, acmc-1awl5, 3-methyl-4-decanol, erythro + threo, 3-methyl-4-decanol, threo + erythro
143312
3-methyldecan-4-ol
CCCCCCC(C(C)CC)O
This item is not returnable. View return policy
55816-17-6
C11H24O
172.312
MFCD00046702
SBMFCRFCWMMVQC-UHFFFAOYSA-N
3-methyl-4-decanol, 4-decanol, 3-methyl, 2-butyl n-hexyl carbinol, acmc-1awl5, 3-methyl-4-decanol, erythro + threo, 3-methyl-4-decanol, threo + erythro
143312
3-methyldecan-4-ol
CCCCCCC(C(C)CC)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 172.31
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-decanol, erythro + threo
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'