Learn More
3-Methyl-3-(methylamino)butanoic Acid Methyl Ester, TRC
Product Code. 30472620
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472620 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30472620 Supplier Toronto Research Chemicals Supplier No. TRCM322035100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Methyl-3-(methylamino)butanoic Acid Methyl Ester
CAS 89855-40-3
Molecular Formula C7H15NO2
Quantity 100 mg
Synonym Methyl 3-methyl-3-(methylamino)butanoate
InChI Formula InChI=1S/C7H15NO2/c1-7(2,8-3)5-6(9)10-4/h8H,5H2,1-4H3
SMILES O=C(OC)CC(NC)(C)C
IUPAC Name methyl 3-methyl-3-(methylamino)butanoate
Molecular Weight (g/mol) 145.2
Formula Weight 145.11