Learn More
3-Methyl-2-chloro-3H-imidazo[4,5-f]quinoline, TRC
Product Code. 30471479
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471479 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30471479 Supplier Toronto Research Chemicals Supplier No. TRCM294452250MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Methyl-2-chloro-3H-imidazo[4,5-f]quinoline
CAS 161087-48-5
Molecular Formula C11 H8 Cl N3
Quantity 250 mg
InChI Formula InChI=1S/C11H8ClN3/c1-15-9-5-4-8-7(3-2-6-13-8)10(9)14-11(15)12/h2-6H,1H3
SMILES Cn1c(Cl)nc2c3cccnc3ccc12
IUPAC Name 2-chloro-3-methylimidazo[4,5-f]quinoline
Molecular Weight (g/mol) 217.65
Formula Weight 217.0407