missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, 99%, Honeywell™

Catalog Number p-7120086
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
563-80-4
C5H10O
86.13
MFCD00008919
SYBYTAAJFKOIEJ-UHFFFAOYSA-N
3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
11251
3-methylbutan-2-one
CC(C)C(C)=O
This item is not returnable. View return policy
563-80-4
C5H10O
86.13
MFCD00008919
SYBYTAAJFKOIEJ-UHFFFAOYSA-N
3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
11251
3-methylbutan-2-one
CC(C)C(C)=O

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Melting Point -92°C
Color Colorless
Density 0.805g/mL at 25°C
Boiling Point 94°C to 96°C
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH3
Quantity 250 mL
UN Number UN2397
Beilstein 1071238
Formula Weight 86.13g/mol
Percent Purity 99%
Chemical Name or Material 3-Methyl-2-butanone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'