Learn More
Thermo Scientific™ 3-Methoxypropylamine, 99%
Product Code. 11491097
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491097 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11491097 Supplier Thermo Scientific™ Supplier No. B24095.AU
 Restricted Item
In Stock
Add to basket
CAS: 5332-73-0 | C4H11NO | 89.14 g/mol
TRUSTED_SUSTAINABILITY
Specifications
CAS 5332-73-0
Melting Point <-70°
Density 0.872
Boiling Point 117-119°
Flash Point 27°(81°F)
Molecular Formula C4H11NO
MDL Number MFCD00014831
Quantity 1000 mL
UN Number UN2734
Synonym 1-Amino-3-methoxypropane;
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
SMILES COCCCN
IUPAC Name 3-methoxypropan-1-amine
Molecular Weight (g/mol) 89.14
Formula Weight 89.14
Percent Purity 99%
Physical Form Liquid