Learn More
3'-Methoxybiphenyl-4-sulfonyl chloride, 95%, Thermo Scientific™
Catalog Number p-4812959
Click to view available options
Quantity:
1 g
5 g
186550-26-5
C13H11ClO3S
282.75
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
10826798
4-(3-methoxyphenyl)benzenesulfonyl chloride
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
186550-26-5
C13H11ClO3S
282.75
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
10826798
4-(3-methoxyphenyl)benzenesulfonyl chloride
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Melting Point 81°C to 83°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 282.75
Percent Purity 95%
Chemical Name or Material 3'-Methoxybiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 3'-Methoxybiphenyl-4-sulfonyl chloride, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.