Learn More
3-Methoxybenzyl alcohol, 97+%
Product Code. p-7022611
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412847 25 g 25g
11422847 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11412847 Supplier Thermo Scientific Alfa Aesar Supplier No. A15471.14
Add to basket
CAS: 6971-51-3 | C8H10O2 | 138.166 g/mol
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxybenzyl alcohol is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in ether, and alcohol. Insoluble in water.
Notes
Incompatible with Oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6971-51-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004637
InChI Key IIGNZLVHOZEOPV-UHFFFAOYSA-N
Synonym 3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol
PubChem CID 81437
ChEBI CHEBI:1580
IUPAC Name (3-methoxyphenyl)methanol
SMILES COC1=CC=CC(=C1)CO
Specifications
Density 1.112
Boiling Point 252°C to 254°C
Flash Point 109°C (228°F)
Refractive Index 1.544
Quantity 25 g
Beilstein 2042063
Solubility Information Soluble in ether,and alcohol. Insoluble in water.
Formula Weight 138.17
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzyl alcohol