missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxyacetophenone, 98%

Product Code. p-156797 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
This item is not returnable. View return policy

Product Code. 10726151

Brand: Thermo Scientific Acros 125680500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 586-37-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-37-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008736
InChI Key BAYUSCHCCGXLAY-UHFFFAOYSA-N
Synonym 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
PubChem CID 11460
IUPAC Name 1-(3-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -7.0°C
Color Brown to Yellow
Density 1.0900g/mL
Boiling Point 239.0°C to 241.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCH3
Refractive Index 1.5394 to 1.5414
Quantity 50 mL
Beilstein 08, 86
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible
Formula Weight 150.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3'-Methoxyacetophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3'-Methoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.