Learn More
3-Methoxy Loratadine, TRC
Product Code. 30484094
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484094 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30484094 Supplier Toronto Research Chemicals Supplier No. TRCM26264010MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Methoxy Loratadine
CAS 165739-73-1
Molecular Formula C23 H25 Cl N2 O3
Quantity 10 mg
InChI Formula InChI=1S/C23H25ClN2O3/c1-3-29-23(27)26-10-8-15(9-11-26)21-20-7-6-18(24)12-16(20)4-5-17-13-19(28-2)14-25-22(17)21/h6-7,12-14H,3-5,8-11H2,1-2H3
SMILES CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4cc(OC)cnc24)CC1
IUPAC Name ethyl 4-(8-chloro-3-methoxy-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
Molecular Weight (g/mol) 412.91
Formula Weight 412.1554