missing translation for 'onlineSavingsMsg'
Learn More

3-Isocyanatobenzoyl chloride, 97%, Thermo Scientific™

Catalog Number 11382847
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5180-79-0
C8H4ClNO2
181.58
MFCD00013864
WBLVOWAHPPPNLF-UHFFFAOYSA-N
acmc-1awgk, 3-isocyanatobenzoylchloride, benzoyl chloride,3-isocyanato, 3-isocyanatobenzoic acid chloride, 3-isocyanatobenzoyl chloride, benzoyl chloride, 3-isocyanato-9ci
5214895
3-isocyanatobenzoyl chloride
ClC(=O)C1=CC=CC(=C1)N=C=O
This item is not returnable. View return policy
5180-79-0
C8H4ClNO2
181.58
MFCD00013864
WBLVOWAHPPPNLF-UHFFFAOYSA-N
acmc-1awgk, 3-isocyanatobenzoylchloride, benzoyl chloride,3-isocyanato, 3-isocyanatobenzoic acid chloride, 3-isocyanatobenzoyl chloride, benzoyl chloride, 3-isocyanato-9ci
5214895
3-isocyanatobenzoyl chloride
ClC(=O)C1=CC=CC(=C1)N=C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.333
Boiling Point 91°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.579
Quantity 1 g
UN Number UN2922
Beilstein 2090273
Sensitivity Moisture sensitive
Formula Weight 181.58
Percent Purity 97%
Chemical Name or Material 3-Isocyanatobenzoyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'