Learn More
3-Isochromanone, Thermo Scientific Chemicals
Product Code. 17231146 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17231146 5 g 5g
17221146 1 g 1g
17241146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17231146 Supplier Thermo Scientific Acros Supplier No. 370130050
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4385-35-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
InChI Key ILHLUZUMRJQEAH-UHFFFAOYSA-N
IUPAC Name 3,4-dihydro-1H-2-benzopyran-3-one
SMILES O=C1CC2=CC=CC=C2CO1
Specifications
Melting Point 83.0°C to 86.0°C
Quantity 5 g
Beilstein 17, V,10, 19
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Formula Weight 148.16
Chemical Name or Material 3-Isochromanone