Learn More
3-Indolyl Acetate, Sigma-Aldrich™
Product Code. 15811131
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15811131 5 g 5g
15801131 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15811131 Supplier Sigma Aldrich Supplier No. 8207060005
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 608-08-2
Grade Research
Molecular Weight (g/mol) 175.18
Specifications
Melting Point 127°C to 129°C
MDL Number MFCD00014561
Quantity 5 g
Synonym 3-Indolyl acetate, Acetic acid 3-indolyl ester, 3-Acetoxyindole, Indoxyl acetate, 3-Indolyl acetate