Learn More
3-Hydroxymethylpiperidine, TRC
Product Code. 30405427
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405427 500 mg 500mg
30390683 2.5 g 2.5g
30404619 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30405427 Supplier Toronto Research Chemicals Supplier No. TRCH972450500MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4606-65-9
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
Synonym 3-Piperidinemethanol, (3-Piperidinyl)methanol, (3-Piperidyl)methanol, (Piperidin-3-yl)methanol, 3-(Hydroxymethyl)piperidine, 3-Pipecolin-α-ol
IUPAC Name piperidin-3-ylmethanol
SMILES OCC1CCCNC1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C6H13NO/c8-5-6-2-1-3-7-4-6/h6-8H,1-5H2
Formula Weight 115.0997
Chemical Name or Material 3-Hydroxymethylpiperidine