Learn More
3-Heptanone, 98%
Product Code. 11428816
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
Product Code. Quantity unitSize
11428816 50 mL 50mL
11438816 250 mL 250mL
11418816 1000 mL 1000mL
3 options
Product Code. 11428816
Brand: Thermo Scientific Alfa Aesar A16977.AD
Request Bulk or Custom Format
CAS: 106-35-4 | C7H14O | 114.19 g/mol
Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.
Solubility
Soluble in water at 20°C 4.3g/L.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-35-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009483
InChI Key NGAZZOYFWWSOGK-UHFFFAOYSA-N
Synonym 3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
PubChem CID 7802
ChEBI CHEBI:50139
IUPAC Name heptan-3-one
SMILES CCCCC(=O)CC
Specifications
Melting Point -37°C
Boiling Point 147°C to 149°C
Flash Point 41°C (105°F)
Odor Pungent
Quantity 50 mL
UN Number UN1224
Beilstein 506161
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 3-Heptanone