Learn More
3-Furaldehyde, TRC
Product Code. 30405619
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405619 5 g
30397501 10 g
30390124 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30405619 Supplier Toronto Research Chemicals Supplier No. TRCF8632005G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 498-60-2
Molecular Formula C5 H4 O2
Molecular Weight (g/mol) 96.08
Synonym 3-Furancarboxaldehyde, 3-Formylfuran, 3-Furanaldehyde, 3-Furancarbaldehyde, 3-Furfural, 3-Furfuraldehyde, 3-Furylaldehyde, 3-Furylcarboxaldehyde
IUPAC Name furan-3-carbaldehyde
SMILES O=Cc1cocc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C5H4O2/c6-3-5-1-2-7-4-5/h1-4H
Formula Weight 96.0211
Chemical Name or Material 3-Furaldehyde