missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Formylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.
Solubility
Slightly soluble in water.
Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.
Solubility
Slightly soluble in water.
Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | ∽180°C (decomposition) |
Quantity | 1 g |
Beilstein | 4387740 |
Sensitivity | Air sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 155.97 |
Percent Purity | 97% |
Chemical Name or Material | 3-Formylthiophene-2-boronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'