Learn More
3-Fluorophthalic acid, 97%
Product Code. 11490244
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11490244
Brand: Thermo Scientific Alfa Aesar A13656.06
Request Bulk or Custom Format
CAS: 1583-67-1 | C8H5FO4 | 184.122 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1583-67-1
Molecular Formula C8H5FO4
Molecular Weight (g/mol) 184.122
MDL Number MFCD00039524
InChI Key BBCQSMSCEJBIRD-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1,2-dicarboxylic acid, 3-fluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylic acid, 3-fluoro, 1,2-benzenedicarboxylic acid, fluoro, 3-fluorophthalic, 3-fluorophtalic acid, 3-fluorophthatic acid, pubchem1405, 3-fluorophthalicacid
PubChem CID 345381
IUPAC Name 3-fluorophthalic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)C(=O)O
Specifications
Melting Point ∼170°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 2616684
Formula Weight 184.12
Percent Purity 97%
Chemical Name or Material 3-Fluorophthalic acid