missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethoxy)phenylacetonitrile, 97%, Thermo Scientific™

Catalog Number 15488675
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1352999-52-0
C9H5F4NO
219.139
MFCD18783227
FISAVDYKHYBCGM-UHFFFAOYSA-N
3-fluoro-5-trifluoromethoxy phenylacetonitrile, 2-3-fluoro-5-trifluoromethoxy phenyl acetonitrile
66523549
2-[3-fluoro-5-(trifluoromethoxy)phenyl]acetonitrile
C1=C(C=C(C=C1OC(F)(F)F)F)CC#N
This item is not returnable. View return policy
1352999-52-0
C9H5F4NO
219.139
MFCD18783227
FISAVDYKHYBCGM-UHFFFAOYSA-N
3-fluoro-5-trifluoromethoxy phenylacetonitrile, 2-3-fluoro-5-trifluoromethoxy phenyl acetonitrile
66523549
2-[3-fluoro-5-(trifluoromethoxy)phenyl]acetonitrile
C1=C(C=C(C=C1OC(F)(F)F)F)CC#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3276
Formula Weight 219.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethoxy)phenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'