Learn More
3-Fluoro-5-methylphenol, 97%, Thermo Scientific™
Catalog Number 11372509
Click to view available options
Quantity:
1 g
5 g
216976-31-7
C7H7FO
126.13
MFCD04115885
AYMBVFCWNCCREA-UHFFFAOYSA-N
phenol, 3-fluoro-5-methyl-9ci, 3-fluoro-5-hydroxytoluene, acmc-1cc83
3549669
3-fluoro-5-methylphenol
CC1=CC(O)=CC(F)=C1
216976-31-7
C7H7FO
126.13
MFCD04115885
AYMBVFCWNCCREA-UHFFFAOYSA-N
phenol, 3-fluoro-5-methyl-9ci, 3-fluoro-5-hydroxytoluene, acmc-1cc83
3549669
3-fluoro-5-methylphenol
CC1=CC(O)=CC(F)=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Formula Weight 126.13
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-methylphenol